Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RKHD3 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281777AF700
This item is not returnable.
View return policy
Description
RKHD3 Polyclonal antibody specifically detects RKHD3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
RKHD3 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
Alexa Fluor 700 | |
DKFZp434J0617, KIAA2009, mex-3 homolog B (C. elegans), ring finger and KH domain containing 3, RING finger and KH domain-containing protein 3, RING finger protein 195, RKHD3MGC117199, RNA-binding protein MEX3B, RNF195MEX-3B | |
RKHD3 antibody was raised against a 16 amino acid synthetic peptide from near the center of human RKHD3. The immunogen is located within amino acids 180 - 230 of RKHD3. Amino Acid Squence: TYIVTPSRDKEPVFEV | |
0.1 mL | |
Zinc Finger | |
84206 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction