Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RKHD4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566520
Description
Immunogen sequence: THIAVRTGKIL EYNNENDFLA GSPDAAIDSR YSDAWRVHQP GCKPLSTFRQ NSLGCI Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.
Rkhd4, also known as MEX3A is a member of a novel family of four homologous human MEX3 proteins each containing two heterogeneous nuclear ribonucleoprotein K homology (KH) domains and one carboxy-terminal RING finger module. MEX3 proteins, including Rkhd4, are phosphoproteins that bind RNA through their KH domains and shuttle between the nucleus and the cytoplasm via the CRM1 export pathway. These proteins are a novel family of evolutionarily conserved RNA-binding proteins, differentially recruited to P bodies and potentially involved in post-transcriptional regulatory mechanisms. While Rkhd2 has been suggested to be associated with susceptibility to essential hypertension type 8, the function of Rkhd4 remains unknown. Rkhd3 and Rkhd4, but not Rkhd2, co-localize with both the hDcp1a decapping factor and Argonaute (Ago) proteins in processing bodies (P bodies), recently characterized as centers of mRNA turnover.
Specifications
RKHD4 | |
Polyclonal | |
Unconjugated | |
MEX3A | |
2700083E18Rik; Gm411; mex3 homolog A; mex3 homolog A (C. elegans); mex-3 RNA binding family member A; MEX3A; MEX-3A; RGD1564904; ring finger and KH domain containing 4; ring finger and KH domain containing protein; RING finger and KH domain-containing protein 4; RKHD4; RNA-binding protein MEX3A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
92312 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A1L020 | |
MEX3A | |
Recombinant Human RKHD4. Recombinant protein control fragment (Product #RP-107187). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction