Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RKI 1447 dihydrochloride
GREENER_CHOICESDP

Catalog No. 506110R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-611-0R 10 mg
50-615-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 50-611-0R Supplier Tocris Bioscience™ Supplier No. 5061/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective ROCK inhibitor; antitumor

RKI 1447 dihydrochloride is a potent and selective ROCK inhibitor (IC50 values are 6.2 and 14.5 nM for ROCK2 and ROCK1 respectively); suppresses phosphorylation of ROCK substrates MLC-2 and MYPT-1 in human cancer cells, but has no effect on the phosphorylation levels of Akt, MEK, and S6 kinase. Displays anti-invasive and antitumor activities in breast cancer cells.

Specifications

CAS 1782109-09-4
Quantity 10 mg
Product Type RKI 1447 dihydrochloride
Purity >99%
Molecular Weight (g/mol) 399.29
Inhibitors RKI 1447 dihydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.