Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RKIP Mouse anti-Human, Unconjugated, Clone: 7F12, MilliporeSigma™

Catalog No. MABC113625 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1136-25 25 μg
MABC1136-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC1136-25 Supplier MilliporeSigma Supplier No. MABC113625UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detects Raf kinase inhibitor protein using this mouse monoclonal Anti-RKIP, clone 7F12, Cat. No. MABC1136, tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RKIP
Applications Western Blot
Classification Monoclonal
Clone 7F12
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in PBS without azide.
Gene Alias Raf kinase inhibitor protein;Phosphatidylethanolamine-binding protein 1;PEBP-1;HCNPpp;Neuropolypeptide h3;Prostatic-binding protein
Gene Symbols PEBP1;PBP;PEBP
Host Species Mouse
Immunogen Purified native and denatured Raf kinase inhibitor protein (RKIP).
Molecular Weight of Antigen ∽23 kDa; 21.06 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_002558.1
Test Specificity Clone 7F12 specifically detects human Raf kinase inhibitor protein, (RKIP).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.