Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RLA-DQ Monoclonal Antibody (2C4), PE
GREENER_CHOICE

Catalog No. PIMA528746
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528746 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528746 Supplier Invitrogen™ Supplier No. MA528746
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RLA-DQ Monoclonal Antibody for Flow

The monoclonal antibody 2C4 reacts with RLA-DQ-transfected cells, but not with RLA-DR-transfected cells. In normal adult rabbits, the antibody reacts with 30-60% of peripheral blood leukocytes. It has also been reported that the antibody stains lymphocytes and macrophages in the spleen, lymphocytes in the thymic medulla and some cells in the cortex, Kupffer cells in the liver, and synovial lining cells (e.g., infiltrating macrophages) in inflamed synovium. DQ alpha transcripts have been detected in bone marrow and spleen and to a lesser extent in appendix, lymph nodes, and other organs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RLA-DQ
Applications Flow Cytometry
Classification Monoclonal
Clone 2C4
Concentration 0.1 mg/mL
Conjugate PE
Formulation PBS with proprietary stabilizer and <0.1% sodium azide; pH 7.5
Gene LOC100343144
Gene Accession No. 0
Gene Alias HLA class II histocompatibility antigen, DQ alpha 1 chain; SLA class II histocompatibility antigen, DQ haplotype D alpha chain; LOC100343144; MHC class II antigen; Orcu-DQA; RLA-DQA; SLA class II histocompatibility antigen, DQ haplotype D alpha chain
Gene Symbols LOC100343144
Host Species Mouse
Immunogen Rabbit spleen cells.
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100343144
Target Species Rabbit
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.