Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RLA-DR Monoclonal Antibody (RDR34)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA528743
Description
RLA-DR Monoclonal Antibody for Flow
The monoclonal antibody RDR34 reacts with RLA-DR-transfected cells but not with RLA-DQ-transfected cells. In normal adult rabbits, the antibody reacts with 50-60% of peripheral blood leukocytes and splenocytes. It has also been reported that the antibody stains lymphocytes in the thymic medulla and some cells in the cortex, periportal cells in the liver, and synovial lining cells (e.g., infiltrating macrophages) in inflamed synovium. DR alpha transcripts have been detected in appendix, bone marrow, spleen, and to a lesser extent in lymph nodes, lung, liver, and other organs.
Specifications
RLA-DR | |
Monoclonal | |
0.5 mg/mL | |
BBS with no preservative; pH 8.2 | |
0 | |
RLA-DR-ALPHA | |
Rabbit leukemic B cell line. | |
1 mL | |
Primary | |
Rabbit | |
Antibody | |
IgG2b κ |
Flow Cytometry | |
RDR34 | |
Unconjugated | |
RLA-DR-ALPHA | |
major histocompatibility complex, class II, DR alpha; RLA-DR-ALPHA | |
Mouse | |
Purified | |
RUO | |
100328929 | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction