Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RLBP1 Monoclonal Antibody (OTI1H5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503185S


Description
RLBP1 Monoclonal antibody specifically detects RLBP1 in Human samples. It is validated for Western BlotSpecifications
RLBP1 | |
Monoclonal | |
0.76 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q15785 | |
Mouse | |
Affinity chromatography | |
RUO | |
10953 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2a |
Western Blot | |
OTI1H5 | |
Unconjugated | |
Tomm34 | |
3110056M11Rik; cellular retinaldehyde-binding protein; cellular retinaldehyde-binding protein-1; CRALBP; retinaldehyde binding protein 1; retinaldehyde-binding protein 1; retinaldehyde-binding protein precursor; Rlbp1 | |
Full length human recombinant protein of human TOMM34 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction