Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RLBP1 Monoclonal Antibody (OTI5G2), TrueMAB™, OriGene
SDP

Catalog No. 502541083
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1083 30 μL
50-167-4118 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1083 Supplier OriGene Supplier No. TA503151S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RLBP1 Monoclonal antibody specifically detects RLBP1 in Human samples. It is validated for Western Blot

Specifications

Antigen RLBP1
Applications Western Blot
Classification Monoclonal
Clone OTI5G2
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RLBP1
Gene Accession No. P12271
Gene Alias 3110056M11Rik; cellular retinaldehyde-binding protein; cellular retinaldehyde-binding protein-1; CRALBP; retinaldehyde binding protein 1; retinaldehyde-binding protein 1; retinaldehyde-binding protein precursor; Rlbp1
Host Species Mouse
Immunogen Full length human recombinant protein of human RLBP1 produced in HEK293 cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6017
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.