Learn More
Invitrogen™ RLF Monoclonal Antibody (2G2)

Description
Peptide Sequence: SNDLTGNVVA NNMVNDSEPE VDIPHSSSDS TIHENLTAIP PLIVAETTTV PSLENLRVVL DKALTDCGEL ALKQLHYLRP VVVLERSKFS TPILDLFPTK KTDELCVGS.
Specifications
Specifications
| Antigen | RLF |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2G2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | RLF |
| Gene Accession No. | Q13129 |
| Gene Alias | rearranged L-myc fusion; rearranged L-myc fusion gene protein; rearranged L-myc fusion sequence; RLF; Zinc finger protein Rlf; Zn-15 related; ZN-15L; zn-15-related protein; ZNF292L |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.