Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RLIM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501730730 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-0730 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-0730 Supplier Proteintech Group Inc Supplier No. 161211AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNF12 is a RING-H2 zinc finger protein. It has been shown to be a ubiquitin protein ligase that targets LIM domain binding 1 (LDB1/CLIM), and causes proteasome-dependent degradation of LDB1. This protein and LDB1 are co-repressors of LHX1/LIM-1, a homeodomain transcription factor. Alternatively spliced transcript variants encoding the same protein have been reported.

Specifications

Antigen RLIM
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RLIM
Gene Accession No. Q9NVW2, Q9WTV7
Gene Alias NY REN 43, R LIM, RING finger protein 12, RLIM, RNF12
Gene Symbols RLIM
Host Species Rabbit
Immunogen RLIM Fusion Protein Ag9077
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108348168, 19820, 51132
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.