Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RLIP76 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501567874
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7874 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7874 Supplier Bethyl Laboratories, Inc Supplier No. A304498A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

May coordinate the cellular actions of activated EGF receptors and Ral-GTPases By similarity. Mungall A.J., Nature 425:805-811(2003). Mural R.J., Submitted (SEP-2005) to the EMBL/GenBank/DDBJ databases. Kimura K., Genome Res. 16:55-65(2006).

Specifications

Antigen RLIP76
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene RALBP1
Gene Accession No. Q15311
Gene Alias Cell migration-inducing gene 2 protein, Raft-linking protein, RFTN1, KIAA0084, ORF Names: MIG2
Gene Symbols RALBP1
Host Species Rabbit
Immunogen Between 605 and 655
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10928
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.