Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RLK/TXK Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$377.03 - $728.30
Specifications
| Antigen | RLK/TXK |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
RLK/TXK Polyclonal specifically detects RLK/TXK in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| RLK/TXK | |
| Polyclonal | |
| Rabbit | |
| Protein Kinase | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 7294 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:REPCNLALRRAEEYLILEKYNPHWWKARDRLGNEGLIP | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| BTKLEC 2.7.10.2, EC 2.7.10, MGC22473, Protein-tyrosine kinase 4, PSCTK5, PTK4, PTK4 protein tyrosine kinase 4, RLK, TKL, TXK tyrosine kinase, tyrosine kinase, tyrosine-protein kinase TXK | |
| TXK | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title