Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RLTPR Antibody (EM-53), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262215AF488
Description
RLTPR Monoclonal specifically detects RLTPR in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
| RLTPR | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RLTPR | |
| Murine Rltpr (amino acids 1147-1397) | |
| 0.1 mL | |
| 146206 | |
| Human, Mouse | |
| IgG1 κ |
| Western Blot, Flow Cytometry | |
| EM-53 | |
| Western Blot, Flow Cytometry | |
| CARMIL2, CARMIL2b, leucine rich repeat containing 16C, leucine-rich repeat-containing protein 16C, LRRC16C, RGD motif, leucine rich repeats, tropomodulin domain and proline-rich containing, RGD, leucine-rich repeat, tropomodulin and proline-rich containing protein, RGD, leucine-rich repeat, tropomodulin and proline-rich-containing protein | |
| Mouse | |
| Protein A purified | |
| Primary | |
| The mouse monoclonal antibody EM-53 recognizes RLTPR/ CARMIL2, a protein playing a role in actin filament elongation. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction