Learn More
RLTPR Antibody (EM-53), DyLight 650, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP262215C
Catalog No. NB002612
Only null left
Add to Cart
Description
Description
RLTPR Monoclonal specifically detects RLTPR in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
Specifications
RLTPR
Monoclonal
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
RLTPR
Murine Rltpr (amino acids 1147-1397)
0.1 mL
146206
Human, Mouse
IgG1 κ
Western Blot, Flow Cytometry
EM-53
Western Blot, Flow Cytometry
CARMIL2, CARMIL2b, leucine rich repeat containing 16C, leucine-rich repeat-containing protein 16C, LRRC16C, RGD motif, leucine rich repeats, tropomodulin domain and proline-rich containing, RGD, leucine-rich repeat, tropomodulin and proline-rich containing protein, RGD, leucine-rich repeat, tropomodulin and proline-rich-containing protein
Mouse
Protein A purified
Primary
The mouse monoclonal antibody EM-53 recognizes RLTPR/ CARMIL2, a protein playing a role in actin filament elongation.
Store at 4°C in the dark.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.