Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RLTPR Antibody (EM-53), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262215IR
Description
RLTPR Monoclonal specifically detects RLTPR in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
RLTPR | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RLTPR | |
Murine Rltpr (amino acids 1147-1397) | |
0.1 mL | |
146206 | |
Human, Mouse | |
IgG1 κ |
Western Blot, Flow Cytometry | |
EM-53 | |
Western Blot, Flow Cytometry | |
CARMIL2, CARMIL2b, leucine rich repeat containing 16C, leucine-rich repeat-containing protein 16C, LRRC16C, RGD motif, leucine rich repeats, tropomodulin domain and proline-rich containing, RGD, leucine-rich repeat, tropomodulin and proline-rich containing protein, RGD, leucine-rich repeat, tropomodulin and proline-rich-containing protein | |
Mouse | |
Protein A purified | |
Primary | |
The mouse monoclonal antibody EM-53 recognizes RLTPR/ CARMIL2, a protein playing a role in actin filament elongation. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction