Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RLTPR Monoclonal Antibody (EM-53)
GREENER_CHOICE

Catalog No. PIMA528632
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528632 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528632 Supplier Invitrogen™ Supplier No. MA528632
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes RLTPR / CARMIL2, an intracellular protein playing a role in actin filament elongation.

RLTPR/CARMIL2 (RGD motif, leucine rich repeats, tropomodulin domain and proline-rich containing; capping protein regulator and myosin 1 linker 2), also known as LRRC16C, is a cytosolic protein, which with high affinity binds CAPZA2 (capping protein muscle actin Z-line alpha 2) and decreases CAPZA2 affinity for actin barbed ends. RLTPR/CARMIL2 increases the rate of actin filament elongation from seeds in the presence of CAPZA2, however, seems unable to nucleate filaments. Its interaction with CAPZA2 is essential for lamellipodial protrusion and cell translocation. RLTPR/CARMIL2 is crucial for T cell costimulation via CD28 and this property seems to be independent on its actin-uncapping function. The lack of functional RLTPR/CARMIL2 molecules impeded the differentiation toward Th1 and Th17 fates of both human and murine CD4+ T cells and leads to combined immunodeficiency. Expression of RLTPR/CARMIL2 was also detected in human and murine B cells, but it seems not to be involved in BCR-mediated signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RLTPR
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone EM-53
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene CARMIL2
Gene Accession No. Q3V3V9, Q6F5E8
Gene Alias Capping protein regulator and myosin 1 linker 2; capping protein, Arp2/3 and myosin-I linker protein 2; CARMIL2; CARMIL2b; CG1399-PB; D130029J02Rik; F-actin-uncapping protein RLTPR; Gm585; leucine rich repeat containing 16C; leucine-rich repeat-containing protein 16C; LRRC16C; RGD motif, leucine rich repeats, tropomodulin domain and proline-rich containing; RGD, leucine-rich repeat, tropomodulin and proline-rich containing protein; RGD, leucine-rich repeat, tropomodulin and proline-rich-containing protein; RLTPR
Gene Symbols CARMIL2
Host Species Mouse
Immunogen Murine Rltpr.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146206, 234695
Target Species Human, Mouse
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.