Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RM30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116684
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116684 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116684 Supplier Invitrogen™ Supplier No. PA5116684
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, U-251MG, 293T Immunogen sequence: MAGILRLVVQ WPPGRLQTVT KGVESLICTD WIRHKFTRSR IPEKVFQASP EDHEKYGGDP QNPHKLHIVT RIKSTRRRPY WEKDIIKMLG LEKAHTPQVH KNIPSVNAKL KVVKHLIRIK PLKLPQGLPA EENMSNTCLK STGELVVQWH LKPVEQKAHE S.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Alternative splicing results in multiple transcript variants. Pseudogenes corresponding to this gene are found on chromosomes 6p and 12p. Read-through transcription also exists between this gene and the neighboring upstream lipoyltransferase 1 (LIPT1) gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RM30
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MRPL30
Gene Accession No. P0C2C1, Q8TCC3
Gene Alias 2310001L22Rik; 39S ribosomal protein L28, mitochondrial; 39S ribosomal protein L30, mitochondrial; HSPC249; L28mt; L30MT; Mitochondrial large ribosomal subunit protein uL30m; mitochondrial ribosomal protein L30; MRPL28; MRP-L28; MRPL28M; MRPL30; MRP-L30; RPML28
Gene Symbols MRPL30
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-161 of human MRPL30 (NP_6602131).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 301352, 51263
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.