Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RMD2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988364
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8364 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8364 Supplier Bioss Supplier No. BS8206R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The human protein FAM82A1, also known as regulator of microtubule dynamics 2 (RMD2), was one of three proteins identified as related to a family of microtubule-associated proteins in C. elegans. FAM82A1 contains multiple coiled-coil domains and localize to the spindle microtubules and spindle poles during cell division. During interphase, RMD2 localizes to the cytoplasm with protein observed in the microtubule lattice and perinuclear region.

Specifications

Antigen RMD2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RMDN2
Gene Accession No. Q96LZ7
Gene Alias AW061290; BLOCK18; FAM82A; FAM82A1; family with sequence similarity 82, member A; family with sequence similarity 82, member A1; microtubule-associated protein; mRMD-2; PRO34163; Protein FAM82A1; PYST9371; regulator of microtubule dynamics 2; Regulator of microtubule dynamics protein 2; RMD2; RMD-2; RMD4; RMDN2; UNQ9371/PRO34163
Gene Symbols RMDN2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FAM82A1/RMD2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151393
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.