Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RMDN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113179
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113179 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113179 Supplier Invitrogen™ Supplier No. PA5113179
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RMDN3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

RMDN3 is a protein coding gene involved in cellular calcium homeostasis regulation. May participate in differentiation and apoptosis of keratinocytes. Overexpression induces apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RMDN3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RMDN3
Gene Accession No. Q3UJU9, Q96TC7
Gene Alias 1200015F23Rik; AI131757; Cerebral protein 10; FAM82A2; Fam82c; family with sequence similarity 82, member A2; family with sequence similarity 82, member C; FLJ10579; hRMD-3; hucep-10; microtubule-associated protein; Protein FAM82A2; Protein FAM82C; Protein tyrosine phosphatase-interacting protein 51; Ptpip51; regulator of microtubule dynamics 3; regulator of microtubule dynamics protein 3; RGD1308697; Rmd3; RMD-3; Rmdn3; TCPTP-interacting protein 51; tyrosine phosphatase interacting protein 51; UNQ3122/PRO10274
Gene Symbols RMDN3
Host Species Rabbit
Immunogen Recombinant Human Regulator of microtubule dynamics protein 3 protein (171-470AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55177, 67809
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.