Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RMDN3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543036
Description
Peptide sequence: TGSSSVYFTA SSGATFTDAE SEGGYTTANA ESDNERDSDK ESEDGEDEVS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
RMDN3 is a protein coding gene involved in cellular calcium homeostasis regulation. May participate in differentiation and apoptosis of keratinocytes. Overexpression induces apoptosis.
Specifications
RMDN3 | |
Polyclonal | |
Unconjugated | |
RMDN3 | |
1200015F23Rik; AI131757; Cerebral protein 10; FAM82A2; Fam82c; family with sequence similarity 82, member A2; family with sequence similarity 82, member C; FLJ10579; hRMD-3; hucep-10; microtubule-associated protein; Protein FAM82A2; Protein FAM82C; Protein tyrosine phosphatase-interacting protein 51; Ptpip51; regulator of microtubule dynamics 3; regulator of microtubule dynamics protein 3; RGD1308697; Rmd3; RMD-3; Rmdn3; TCPTP-interacting protein 51; tyrosine phosphatase interacting protein 51; UNQ3122/PRO10274 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
55177 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96TC7 | |
RMDN3 | |
Synthetic peptide directed towards the N-terminal of human RMDN3 (aa 152-201). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction