Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ RME-8, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABN1657 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1657 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1657 Supplier MilliporeSigma™ Supplier No. ABN1657
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DnaJ homolog subfamily C member 13 (UniProt O75165; also known as Required for receptor-mediated endocytosis 8, RME-8) is encoded by the DNAJC13 (also known as KIAA0678, PARK21, RME8) gene (Gene ID 23317) in human. Originally identified as a C. elegans protein involved in mediating endocytosis, , receptor-mediated endocytosis 8 (RME-8) is a member of the DNAJ family of proteins implicated in regulating protein folding through associated chaperones. RME-8 is shown to interact with Hsc70/HSP-1 and sorting nexin 1 (SNX-1), loss of RME-8, SNX-1 or Hsc70 affects retrograde trafficking at the early endosomes-TGN interface. In addition, RME-8 is demonstrated to coordinate the function of the WASH complex and the retromer SNX dimer. The pentameric WASH complex is comprised of WASH1, strumpellin, SWIP (strumpellin- and wash-interacting protein), FAM21, and CCDC53. RME-8 N-terminal J region (a.a. 1-453) involved in membrane association is found to be involved in establishing interaction with the first third of FAM21 tail domain. WASH complex is responsible for the generation of branched actin networks on endosomes and the creation of microdomains into which specific cargoes can be concentrated. Loss of RME-8 function results in a profound endosomal tubulation, more extensive than the endosomal tubulation phenotype resulted from loss of the WASH complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RME-8
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing PBS with 0.05% Sodium Azide and 50% glycerol.
Gene DNAJC13, KIAA0678, PARK21, RME8
Gene Accession No. O75165
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the C-terminal end sequence of human RME-8.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_056083
Test Specificity This rabbit polyclonal antibody detected shRNA-mediated cellular RME-8 knockwon by immunoblotting and fluorescent immunocytochemistry (Freeman, C.L., et al. (2014). J. Cell Sci. 127(Pt 9):2053-2070; Girard, M., and McPherson, P.S. (2008). FEBS Lett. 582(6):961-966; Girard, M., et al. J. Biol. Chem. 280(48):40135-40143).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.