Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RMI1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110300
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110300 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110300 Supplier Invitrogen™ Supplier No. PA5110300
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNVTSIALRA ETWLLAAWHV KVPPMWLEAC INWIQEENNN VNLSQAQMNK QVFEQWLLTD LRDLEHPLLP DGILEIPKGE LNGFYALQIN SLVDVSQPAY SQIQKLRGKN TTNDLVTAEA QVTPKPWEAK PSRMLMLQLT DGIVQIQGME YQPIPILHSD LPPGTKILIY GNISFRLGVL LLKPENVKVL GGEVDALLEE YAQEKVLARL IGEPDLVVSV IPNNSNENIP RVTDVLDPAL GPSDEELLAS LDENDELTAN NDTSSERCFT TGSSSNTIPT RQSSFEPEFV ISPRPKEEPS NLSIHVMDGE LDDFSLEEAL LLEETVQKEQ METKELQPLT FNRNADRSIE.

Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RMI1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RMI1
Gene Accession No. Q9D4G9, Q9H9A7
Gene Alias 4932432N11Rik; BLAP75; BLM-associated polypeptide, 75 kDa; BLM-associated protein 75 kDa; BLM-associated protein of 75 kDa; C79893; C9orf76; FAAP75; FLJ12888; homolog of yeast RecQ-mediated genome instability 1 (RMI1); RecQ mediated genome instability 1; recQ-mediated genome instability protein 1; recQ-mediated genome instability protein-like protein; RGD1310671; RMI1; RMI1, RecQ mediated genome instability 1, homolog; RMI1, RecQ mediated genome instability 1, homolog (S. cerevisiae); RP11-346I8.1
Gene Symbols RMI1
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74386, 80010
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.