Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RMI1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113411
Description
RMI1 Polyclonal Antibody for Western Blot, ELISA
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The STE group (homologs of yeast Sterile 7, 11, 20 kinases) consists of 50 kinases related to the mitogen-activated protein kinase (MAPK) cascade families (Ste7/MAP2K, Ste11/MAP3K, and Ste20/MAP4K). MAP kinase cascades, consisting of a MAPK and one or more upstream regulatory kinases (MAPKKs) have been best characterized in the yeast pheromone response pathway. Pheromones bind to Ste cell surface receptors and activate yeast MAPK pathway.
Specifications
RMI1 | |
Polyclonal | |
Unconjugated | |
RMI1 | |
4932432N11Rik; BLAP75; BLM-associated polypeptide, 75 kDa; BLM-associated protein 75 kDa; BLM-associated protein of 75 kDa; C79893; C9orf76; FAAP75; FLJ12888; homolog of yeast RecQ-mediated genome instability 1 (RMI1); RecQ mediated genome instability 1; recQ-mediated genome instability protein 1; recQ-mediated genome instability protein-like protein; RGD1310671; RMI1; RMI1, RecQ mediated genome instability 1, homolog; RMI1, RecQ mediated genome instability 1, homolog (S. cerevisiae); RP11-346I8.1 | |
Rabbit | |
Protein G | |
RUO | |
74386, 80010 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9D4G9, Q9H9A7 | |
RMI1 | |
Recombinant Human RecQ-mediated genome instability protein 1 protein (181-443AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction