Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RMI2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP18996225UL
Description
RMI2 Polyclonal specifically detects RMI2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RMI2 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
BLAP18RMI2, BLM-associated protein of 18 kDa, chromosome 16 open reading frame 75, hRMI2, MGC24665, RecQ-mediated genome instability 2, S. cerevisiae, homolog of, recQ-mediated genome instability protein 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
116028 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.3mg/mL | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Q96E14 | |
RMI2 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GKYVMVMGVVQACSPEPCLQAVKMTDLSDNPIHESMWELEVEDLHRNI | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction