Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RMI2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597103
Description
Immunogen sequence: MAAAADSFSG GPAGVRLPRS PPLKVLAEQL RRDAEGGPGA WRLSRAAAGR GPLDLAAVWM QGRVVMADRG EARLRDPSGD FSVRGLERVP RGRPCLVPGK YVMVMGVVQA CSPEPCLQAV KMTDLSDNPI HESMWELEVE DLHRNIP; Positive Samples: HeLa, MCF7; Cellular Location: Nucleus.
RMI2 is a protein coding gene, essential component of the RMI complex, a complex that plays an important role in the processing of homologous recombination intermediates. It is required to regulate sister chromatid segregation and to limit DNA crossover. Essential for the stability, localization, and function of BLM, TOP3A, and complexes containing BLM. In the RMI complex, it is required to target BLM to chromatin and stress-induced nuclear foci and mitotic phosphorylation of BLM.
Specifications
RMI2 | |
Polyclonal | |
Unconjugated | |
RMI2 | |
A630055G03Rik; BLAP18; BLM-associated protein of 18 kDa; C16orf75; Gm537; Gm929; hRMI2; RecQ mediated genome instability 2; recQ-mediated genome instability protein 2; RGD1563163; RMI2; RMI2, RecQ mediated genome instability 2, homolog; RMI2, RecQ mediated genome instability 2, homolog (S. cerevisiae); similar to hypothetical protein MGC24665 | |
Rabbit | |
Affinity chromatography | |
RUO | |
116028 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.69 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96E14 | |
RMI2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-147 of human RMI2 (NP_6895211). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction