Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RMI2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559381
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559381 Supplier Invitrogen™ Supplier No. PA559381
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GKYVMVMGVV QACSPEPCLQ AVKMTDLSDN PIHESMWELE VEDLHRNI Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 96%.

RMI2 is a protein coding gene, essential component of the RMI complex, a complex that plays an important role in the processing of homologous recombination intermediates. It is required to regulate sister chromatid segregation and to limit DNA crossover. Essential for the stability, localization, and function of BLM, TOP3A, and complexes containing BLM. In the RMI complex, it is required to target BLM to chromatin and stress-induced nuclear foci and mitotic phosphorylation of BLM.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RMI2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RMI2
Gene Accession No. Q96E14
Gene Alias A630055G03Rik; BLAP18; BLM-associated protein of 18 kDa; C16orf75; Gm537; Gm929; hRMI2; RecQ mediated genome instability 2; recQ-mediated genome instability protein 2; RGD1563163; RMI2; RMI2, RecQ mediated genome instability 2, homolog; RMI2, RecQ mediated genome instability 2, homolog (S. cerevisiae); similar to hypothetical protein MGC24665
Gene Symbols RMI2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RMI2. Recombinant protein control fragment (Product #RP-99922).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116028
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.