Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RMND1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556730
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556730 Supplier Invitrogen™ Supplier No. PA556730
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LKAGKKVKLS HEEVMQKIGE LFALRHRINL SSDFLITPDF YWDRENLEGL YDKTCQFLSI GRRVKVMNEK LQHCMEL Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RMND1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RMND1
Gene Accession No. Q9NWS8
Gene Alias 0610042C05Rik; AA408137; AI462664; AW536662; bA351K16; bA351K16.3; C6orf96; COXPD11; Iag-1; implantation-associated gene-1; required for meiotic nuclear division 1 homolog; required for meiotic nuclear division 1 homolog (S. cerevisiae); required for meiotic nuclear division protein 1 homolog; RGD1309546; RMD1; Rmnd1
Gene Symbols RMND1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RMND1. Recombinant protein control fragment (Product #RP-94914).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55005
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.