Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RMND1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598968
Description
RMND1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
The function of this protein remains unknown.
Specifications
RMND1 | |
Polyclonal | |
Unconjugated | |
RMND1 | |
0610042C05Rik; AA408137; AI462664; AW536662; bA351K16; bA351K16.3; C6orf96; COXPD11; Iag-1; implantation-associated gene-1; required for meiotic nuclear division 1 homolog; required for meiotic nuclear division 1 homolog (S. cerevisiae); required for meiotic nuclear division protein 1 homolog; RGD1309546; RMD1; Rmnd1 | |
Rabbit | |
Protein G | |
RUO | |
55005, 66084 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8CI78, Q9NWS8 | |
RMND1 | |
Recombinant Human Required for meiotic nuclear division protein 1 homolog protein (35-160AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction