Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RMND5A Antibody (OTI2C8), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245606
Description
Ensure accurate, reproducible results in Western Blot
RMND5A Monoclonal specifically detects RMND5A in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
RMND5A | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000 | |
C-terminal to LisH motif, 44 kDa, CTLH, FLJ12753, FLJ13910, FLJ21795, MGC78451, p44CTLH, protein RMD5 homolog A, required for meiotic nuclear division 5 homolog A (S. cerevisiae), RMD5 | |
Mouse | |
43.8 kDa | |
0.1 mL | |
Stem Cell Markers | |
64795 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot | |
OTI2C8 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
RMND5A | |
Human recombinant protein fragment corresponding to amino acids 1-249 of human RMND5A (NP_073617) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction