Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RMND5A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584471
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584471 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584471 Supplier Invitrogen™ Supplier No. PA584471
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RELEKVLHKF SGYGQLCERG LEELIDYTGG LKHEILQSHG QDAELSGTLS LVLTQCCKRI KDT Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

RMND5A is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RMND5A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RMND5A
Gene Accession No. Q9H871
Gene Alias 1110007A06Rik; 44-kD protein coding for CTLH motif; AJ237586; AW125795; C-terminal to LisH motif, 44 kDa; CTLH; E3 ubiquitin-protein ligase RMND5A; E3 ubiquitin-protein transferase RMND5A; FLJ13910; GID complex subunit 2 homolog A; GID2; GID2A; MTA.D02.090; p44CTLH; Protein RMD5 homolog A; required for meiotic nuclear division 5 homolog A; required for meiotic nuclear division 5 homolog A (S. cerevisiae); RGD1309766; RMD5; RMND5A
Gene Symbols RMND5A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RMND5A. Recombinant protein control fragment (Product #RP-105159).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64795
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.