Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RN 1 dihydrochloride
GREENER_CHOICE

Catalog No. 497710
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-771-0 10 mg
49-775-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 49-771-0 Supplier Tocris Bioscience™ Supplier No. 4977/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

LSD1 inhibitor

Potent lysine specific demethylase 1 (LSD1) inhibitor (IC50 = 70 nM). Exhibits selectivity for LSD1 over MAO-A and MAO-B (IC50 values are 0.51 and 2.79 μM for MAO-A and MAO-B, respectively). Impairs long-term, but not short-term memory in mice. Brain penetrant following systemic administration.

Chemical Identifiers

Molecular Formula C23H29N3O2·2HCl

Specifications

Quantity 10 mg
Formula Weight 452.42
Chemical Name or Material RN 1 dihydrochloride
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.