Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RN 1747, Tocris Bioscience™

Catalog No. 374510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-451-0 10 mg
37-455-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-451-0 Supplier R&D Systems Supplier No. 3745/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Selective TRPV4 agonist

Selective TRPV4 agonist (EC50 values are 0.77, 4.0 and 4.1 μM for hTRPV4, mTRPV4 and rTRPV4 respectively). Displays selectivity over other TRP channels (EC50 values are 0.77, >30, >30 and >100 μM for TRPV4, TRPM8, TRPV3 and TRPV1 receptors respectively). Also antagonizes TRPM8 at relevant concentrations (IC50 = 4 μM).

Chemical Identifiers

CAS 1024448-59-6
Molecular Formula C17H18ClN3O4S
Molecular Weight (g/mol) 395.858
InChI Key ZNLVYSJQUMALEO-UHFFFAOYSA-N
Synonym 4-chloro-2-nitro-1-4-benzylpiperazinyl sulfonyl benzene, 1-benzyl-4-4-chloro-2-nitrobenzenesulfonyl piperazine, d0a5ly, rn hplc, 1-4-chloro-2-nitrophenyl sulfonyl-4-benzylpiperazine, 1-benzyl-4-4-chloro-2-nitrophenyl sulfonylpiperazine, 1-4-chloro-2-nitrophenyl sulfonyl-4-benzylpiperazine;, 1-benzyl-4-4-chloro-2-nitrobenzene sulfonyl piperazine, 1-4-chloro-2-nitrophenyl sulfonyl-4-phenylmethyl-piperazine
PubChem CID 5068295
IUPAC Name 1-benzyl-4-(4-chloro-2-nitrophenyl)sulfonylpiperazine
SMILES C1CN(CCN1CC2=CC=CC=C2)S(=O)(=O)C3=C(C=C(C=C3)Cl)[N+](=O)[O-]

Specifications

Quantity 10 mg
Formula Weight 395.87
Percent Purity >98%
Chemical Name or Material RN 1747
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.