Learn More
Cytiva RNA Homopolymers
GSA_VA
Catalog No. 45500119 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 mg
Catalog No. Quantity
45-500-119 500 mg
1 options
Catalog No. 45-500-119 Supplier Cytiva Supplier No. 27411002
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Template primers
These products are dispensed by weight of dry material, which includes polymer, residual salt, and residual water; content of polymer may vary from lot to lot.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Product Type RNA Homopolymer
Content And Storage -20°C
Primer Poly(rA)
Quantity 500 mg