Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva RNA Homopolymers

Catalog No. 45001356 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-356 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-356 Supplier Cytiva Supplier No. 27411001
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Template primers

These products are dispensed by weight of dry material, which includes polymer, residual salt, and residual water; content of polymer may vary from lot to lot.

TRUSTED_SUSTAINABILITY

Specifications

Product Type RNA Homopolymer
Content And Storage -20°C
Primer Poly(rA)
Quantity 100 mg
For Use With (Application) Evaluate the binding of potential therapeutic agents to mRNA in drug development research
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.