Learn More
R&D Systems™ RNA Imaging Probe 1c
SDP
Catalog No. 881350U Shop All Novus Products
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
88-135-0U 50 μg
1 options
This item is not returnable. View return policy
Catalog No. 88-135-0U Supplier R&D Systems™ Supplier No. 8813/50U
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
A Tocris product, part of R&D Systems. Fluorogenic RNA imaging probe
RNA Imaging Probe 1c is a fluorogenic RNA imaging probe. RNA Imaging Probe 1c displays high membrane permeability, strong fluorogenic responses upon binding RNA, compatibility with fluorescence lifetime imaging microscopy (FLIM), low cytotoxicity, and excellent photostability. Excitation and emission maxima (λ) are 556 nm and 608 nm, respectively; quantum yield = 0.49; extinction coefficient = 27,500 M-1cm-1. Suitable for live cell imaging.
Specifications
Quantity 50 μg
Product Line Tocris