Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNA Polymerase II/POLR2A, p Thr4 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149546G
Description
RNA Polymerase II/POLR2A Polyclonal specifically detects RNA Polymerase II/POLR2A in Human, Chicken, Drosophila, Yeast samples. It is validated for Western Blot.Specifications
RNA Polymerase II/POLR2A (p Thr4) | |
Polyclonal | |
Western Blot | |
P24928 | |
POLR2A | |
A synthetic peptide made to a phosphorylated Threonine (position 4) of the CTD heptad in the human RNA polymerase II protein [UniProt P24928] | |
0.1 mL | |
Chromatin Research, Epigenetics, microRNA, Phospho Specific, Transcription Factors and Regulators | |
5430 | |
Human, Chicken, Drosophila, Yeast | |
IgG |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is specific for the phosphorylated Threonine found in position 4 of the RNA polymerase II CTD heptad. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction