Learn More
Invitrogen™ RNA polymerase sigma 70 Polyclonal Antibody

Description
RNA polymerase sigma 70 Polyclonal Antibody for Western Blot, ELISA
Specifications
Specifications
| Antigen | RNA polymerase sigma 70 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 3.43 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 |
| Gene | rpoD |
| Gene Accession No. | P00579 |
| Gene Alias | RNA polymerase sigma factor RpoD (Sigma-70) rpoD alt b3067 JW3039 |
| Gene Symbols | rpoD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.