Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNAse H2A Polyclonal Antibody
RNAse H2A Polyclonal Antibody
Supplier: Thermo Scientific PA590243
Description
RNAse H2A Polyclonal Antibody for Western Blot, ICC/IF

Specifications
RNAse H2A | |
Polyclonal | |
Unconjugated | |
RNASEH2A | |
2400006P09Rik; AGS4; Aicardi-Goutieres syndrome 4 protein; hypothetical protein LOC512830; hypothetical protein MGC56307; JUNB; ribonuclease H2 subunit A; ribonuclease H2, large subunit; ribonuclease H2, subunit A; ribonuclease HI; ribonuclease HI large subunit; Ribonuclease HI subunit A; ribonuclease HI, large subunit; RNase H(35); RNase H2 subunit A; RNase HI large subunit; Rnaseh2a; RNASEHI; RNHIA; RNHL; zgc:56307 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10535, 364974 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.57 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O75792, Q5U209 | |
RNASEH2A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-299 of human RNASEH2A (NP_0063882). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction