Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNase L Antibody (2E9), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100351AF405
Description
RNase L Monoclonal specifically detects RNase L in Human, Hamster, Primate, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RNase L | |
Monoclonal | |
Alexa Fluor 405 | |
Q05823 | |
RNASEL | |
Purified full-length recombinant human RNaseL produced in insect cells. [UniProt# Q05823] | |
Protein G purified | |
RUO | |
Primary | |
Human, hamster and primate. This antibody does not react with mouse or rat. | |
Store at 4C in the dark. | |
IgG1 κ |
Immunoassay, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
2000000000 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
2-5A-dependent ribonuclease, 2-5A-dependent RNase, 2'-5'-oligoisoadenylate synthetase-dependent, EC 3.1.26.-, HPC1, interferon-induced 2-5A-dependent RNase, MGC104972, MGC133329, PRCA1, prostate cancer 1, Ribonuclease 4, Ribonuclease L, ribonuclease L (2'-5'-oligoisoadenylate synthetase-dependent), RNase L, RNS4DKFZp781D08126 | |
Mouse | |
83 kDa | |
0.1 mL | |
Chromatin Research, Cytokine Research, DNA replication Transcription Translation and Splicing, Immunology, Innate Immunity, Prostate Cancer, Protein Kinase | |
6041 | |
Human, Hamster, Primate, Mouse (Negative), Rat (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction