Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNase L Antibody (2E9) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB014002 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014002 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB014002 Supplier Novus Biologicals Supplier No. NBP280929
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RNase L Monoclonal specifically detects RNase L in Human, Hamster, Primate, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RNase L
Applications Immunoassay, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2000000000
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:4000, ELISA : 1:100-1:2000, Immunohistochemistry : 1:200-1:1000, Immunohistochemistry-Paraffin : 1:200-1:1000, Knockout Validated
Formulation PBS
Gene Alias 2-5A-dependent ribonuclease, 2-5A-dependent RNase, 2'-5'-oligoisoadenylate synthetase-dependent, EC 3.1.26.-, HPC1, interferon-induced 2-5A-dependent RNase, MGC104972, MGC133329, PRCA1, prostate cancer 1, Ribonuclease 4, Ribonuclease L, ribonuclease L (2'-5'-oligoisoadenylate synthetase-dependent), RNase L, RNS4DKFZp781D08126
Gene Symbols RNASEL
Host Species Mouse
Immunogen Purified full-length recombinant human RNaseL produced in insect cells. [UniProt# Q05823]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Cytokine Research, DNA replication Transcription Translation and Splicing, Immunology, Innate Immunity, Prostate Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 6041
Test Specificity Human, hamster and primate. This antibody does not react with mouse or rat.
Target Species Human, Hamster, Primate, Mouse (Negative), Rat (Negative)
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.