Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNase L Antibody (2E9), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100351AF750
Description
RNase L Monoclonal specifically detects RNase L in Human, Hamster, Primate, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RNase L | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
RNASEL | |
Purified full-length recombinant human RNaseL produced in insect cells. [UniProtNo. Q05823] | |
0.1 mL | |
Primary | |
Human, Hamster, Primate, Mouse, Rat | |
IgG1 κ |
Immunoassay, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
2000000000 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
2-5A-dependent ribonuclease, 2-5A-dependent RNase, 2′-5′-oligoisoadenylate synthetase-dependent, EC 3.1.26.-, HPC1, interferon-induced 2-5A-dependent RNase, MGC104972, MGC133329, PRCA1, prostate cancer 1, Ribonuclease 4, Ribonuclease L, ribonuclease L (2′-5′-oligoisoadenylate synthetase-dependent), RNase L, RNS4DKFZp781D08126 | |
Mouse | |
Protein G purified | |
Chromatin Research, Cytokine Research, DNA replication Transcription Translation and Splicing, Immunology, Innate Immunity, Prostate Cancer, Protein Kinase | |
6041.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction