Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNase1 Monoclonal Antibody (02)
GREENER_CHOICE

Catalog No. PIMA530818
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530818 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530818 Supplier Invitrogen™ Supplier No. MA530818
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human RNase A/Ribonuclease A/RNASE1.

Endonuclease that catalyzes the cleavage of RNA on the 3' side of pyrimidine nucleotides. Acts on single stranded and double stranded RNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNase1
Applications ELISA
Classification Monoclonal
Clone 02
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 20mM MES with 150mM NaCl, 10% trehalose, 0.02% Tween 80 and no preservative; pH 6.0
Gene RNASE1
Gene Accession No. P07998
Gene Alias AI574248; HP-RNase; RAC1; Rib 1; RIB1; Rib-1; Ribonuclease 1; ribonuclease 1, pancreatic; ribonuclease A; ribonuclease A (pancreatic); ribonuclease A C1; ribonuclease A family member 1, pancreatic; ribonuclease BS-1; Ribonuclease pancreatic; ribonuclease, RNase A family, 1 (pancreatic); RNase 1; RNase A; RNase upI-1; RNASE1; RNAseA; RNS1; Seminal ribonuclease; seminal RNase; seminal RNase (aa 47-124); SRN; S-RNase
Gene Symbols RNASE1
Host Species Mouse
Immunogen Recombinant Human RNase A/Ribonuclease A/RNASE1 protein (Met1-Thr156).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6035
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.