Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNase1 Polyclonal Antibody

RNase1 Polyclonal Antibody
Supplier: Thermo Scientific PA551519
Description
RNase1 Polyclonal Antibody for Western Blot, ICC/IF, IHC

Specifications
RNase1 | |
Polyclonal | |
Unconjugated | |
RNASE1 | |
AI574248; HP-RNase; RAC1; Rib 1; RIB1; Rib-1; Ribonuclease 1; ribonuclease 1, pancreatic; ribonuclease A; ribonuclease A (pancreatic); ribonuclease A C1; ribonuclease A family member 1, pancreatic; ribonuclease BS-1; Ribonuclease pancreatic; ribonuclease, RNase A family, 1 (pancreatic); RNase 1; RNase A; RNase upI-1; RNASE1; RNAseA; RNS1; Seminal ribonuclease; seminal RNase; seminal RNase (aa 47-124); SRN; S-RNase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6035 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P07998 | |
RNASE1 | |
Recombinant protein corresponding to Human RNase1. Recombinant protein control fragment (Product #RP-88876). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction