Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNASE11 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen RNASE11
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP157953
SDP
View Documents
Novus Biologicals
NBP157953
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

RNASE11 Polyclonal specifically detects RNASE11 in Human samples. It is validated for Western Blot.
Specifications

Specifications

RNASE11
Polyclonal
Rabbit
Q8TAA1
122651
Synthetic peptides corresponding to RNASE11(ribonuclease, RNase A family, 11 (non-active)) The peptide sequence was selected from the middle region of RNASE11. Peptide sequence GISCCESLELENTVCQFTTGKQFPRCQYHSVTSLEKILTVLTGHSLMSWL.
Primary
Western Blot
Unconjugated
RUO
C14orf6, chromosome 14 open reading frame 6, EC 3.1.27.-, probable ribonuclease 11, ribonuclease 11, ribonuclease, RNase A family, 11 (non-active), RNase 11
RNASE11
IgG
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.