Learn More
RNASE11 Mouse anti-Human, Clone: OTI1D10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810563

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
RNASE11 (Ribonuclease A Family Member 11 (Inactive)) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and ribonuclease activity.Specifications
RNASE11 | |
Monoclonal | |
Unconjugated | |
RNASE11 | |
C14orf6, HEL-S-84p, RAJ1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
122651 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1D10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8TAA1 | |
RNASE11 | |
Full length human recombit protein of human RNASE11 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.