Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNase11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102350
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102350 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102350 Supplier Invitrogen™ Supplier No. PA5102350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RNASE11.

RNASE11 (Ribonuclease A Family Member 11 (Inactive)) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and ribonuclease activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNase11
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RNASE11
Gene Accession No. Q5GAM9, Q8TAA1
Gene Alias C14orf6; epididymis secretory sperm binding protein Li 84p; HEL-S-84p; Probable ribonuclease 11; putative inactive ribonuclease 11; RAJ1; ribonuclease 11; ribonuclease A family member 11 (inactive); ribonuclease A j1; ribonuclease, RNase A family, 11 (non-active); RNase 11; RNASE11; UNQ5832/PRO19669
Gene Symbols RNASE11
Host Species Rabbit
Immunogen A synthesized peptide derived from human RNASE11(Accession Q8TAA1), corresponding to amino acid residues S65-K115.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122651, 497113
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.