Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNase12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537411
Description
This antibody detects endogenous protein at a molecular weight of 17 kDa. Purity is >95% by SDS-PAGE.
Probable inactive ribonuclease-like protein 12, RNASE 12, belongs to the pancreatic ribonuclease family and does not exhibit any ribonuclease activity.
Specifications
RNase12 | |
Polyclonal | |
Unconjugated | |
RNASE12 | |
epididymis secretory sperm binding protein Li 85p; HEL-S-85p; Probable inactive ribonuclease-like protein 12; Rai1; RGD1307829; ribonuclease 12; ribonuclease A family member 12 (inactive); ribonuclease A I1; ribonuclease, RNase A family, 12 (non-active); ribonuclease-like protein 12; RNASE12; RNS12 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
493901 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q5GAN4 | |
RNASE12 | |
Recombinant full length human RNASE12. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction