Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNASE13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102340
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102340 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102340 Supplier Invitrogen™ Supplier No. PA5102340
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RNASE13.

Does not exhibit any ribonuclease activity. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNASE13
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RNASE13
Gene Accession No. Q5GAL7, Q5GAM7, Q5GAN3
Gene Alias epididymis secretory sperm binding protein Li 86p; HEL-S-86p; pancreatic ribonuclease; probable inactive ribonuclease-like protein 13; RAL1; ribonuclease 13; ribonuclease A family member 13 (inactive); ribonuclease A L1; ribonuclease, RNase A family, 13 (non-active); ribonuclease-like protein 13; Rnase13
Gene Symbols RNASE13
Host Species Rabbit
Immunogen A synthesized peptide derived from human RNASE13(Accession Q5GAN3), corresponding to amino acid residues C68-Q118.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440163, 497071, 497194
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.