Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNASE2 Monoclonal Antibody (C13)
Mouse Monoclonal Antibody
$243.00 - $552.50
Specifications
Antigen | RNASE2 |
---|---|
Clone | C13 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541672
|
Invitrogen™
MA541672 |
200 μL |
Each for $552.50
|
|
|||||
PIMA543605
|
Invitrogen™
MA543605 |
20 μL |
Each for $243.00
|
|
|||||
Description
RNASE2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This is a non-secretory ribonuclease. It is a pyrimidine specific nuclease with a slight preference for U. Cytotoxin and helminthotoxin. Selectively chemotactic for dendritic cells. Possesses a wide variety of biological activities.Specifications
RNASE2 | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P10153 | |
6036 | |
Recombinant protein Ribonuclease A2. The antigen corresponds to amino acid range 28-161 of the target protein. | |
Antibody |
C13 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2a κ | |
Human | |
RNASE2 | |
EDN; eosinophil-derived neurotoxin; Non-secretory ribonuclease; RAF3; Ribonuclease 2; ribonuclease A F3; ribonuclease A family member 2; ribonuclease US; ribonuclease, RNase A family, 2 (liver, eosinophil-derived neurotoxin); RNase 2; RNase UpI-2; RNASE2; RNS2 | |
RNASE2 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title