Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNASE3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120006
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120006 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120006 Supplier Invitrogen™ Supplier No. PA5120006
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: U937 cell lysates, human spleen tissue, human kidney tissue. Subcellular Location: Secreted.

ECP (eosinophil cationic protein), also known as RNS3 or RNASE3 (ribonuclease, RNase A family, 3), is a 160 amino acid potent cytotoxic secretory protein that belongs to the pancreatic ribonuclease family. ECP localizes to the granule matrix of the eosinophil and is involved in the immune response system. ECP is a cytotoxin, neurotoxin and helminthotoxin that is secreted by activated human eosinophils. ECP exhibits anti-parasitic, antibacterial and ribonucleolytic activities. It has been suggested that ECP induces the neurotoxic effect known as the Gordon phenomenon, a syndrome manifested by ataxia, muscular rigidity, paralysis and tremor that may lead to death. ECP is considered a marker of eosinophilic inflammation and high levels have been found in cases of active asthma and other allergic diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNASE3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene RNASE3
Gene Accession No. P12724
Gene Alias cytotoxic ribonuclease; Ear11; EAR-11; Ear3; ECP; Eosinophil cationic protein; eosinophil cationic protein-like; eosinophil cationic-type ribonuclease 3; Eosinophil secondary granule ribonuclease 11; eosinophil-associated ribonuclease 3; eosinophil-associated, ribonuclease A family, member 3; LOC100361866; mR3; MR-3; murine ribonuclease 3; RAF1; Ribonuclease 3; ribonuclease A family member 3; ribonuclease, RNase A family 3; ribonuclease, RNase A family, 3; RNase 3; RNASE3; RNS3
Gene Symbols RNASE3
Host Species Rabbit
Immunogen Recombinant protein within human ribonuclease 3 aa 28-160.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6037
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.